Ethyl 2-(3-(methylthio)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82503-500MG 82503-1G 82503-5G 82503-25G
77334-692EA 96.82 USD
77334-692 77342-650 77370-716 77382-816
Ethyl 2-(3-(methylthio)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-(methylthio)phenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃S
MW: 224.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113526
CAS Number: 1256479-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR