Ethyl-2-isopropylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-isopropylphenyl)-2-oxoacetate

82480-500MG 82480-1G 82480-5G 82480-25G
77334-690EA 199.82 USD
77334-690 77358-880 77370-714 77391-352
Ethyl-2-isopropylbenzoylformate ≥95%
Ethyl-2-isopropylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801428
CAS Number: 951888-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR