Ethyl-2-chloro-4-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-chloro-4-fluorophenyl)-2-oxoacetate

82475-500MG 82475-1G 82475-5G 82475-25G
77334-688EA 199.82 USD
77334-688 77358-878 77370-712 77391-350
Ethyl-2-chloro-4-fluorobenzoylformate ≥95%
Ethyl-2-chloro-4-fluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD09801393
CAS Number: 951887-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR