Ethyl-2,6-difluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2,6-difluorophenyl)-2-oxoacetate

82471-500MG 82471-1G 82471-5G 82471-25G
77334-686EA 199.82 USD
77334-686 77358-876 77370-710 77391-348
Ethyl-2,6-difluorobenzoylformate ≥95%
Ethyl-2,6-difluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09965862
CAS Number: 1049030-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR