Ethyl-3-chloro-4-methylbenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-chloro-4-methylphenyl)-2-oxoacetate

82467-500MG 82467-1G 82467-5G 82467-25G
77334-684EA 95.79 USD
77334-684 77342-636 77370-708 77382-814
Ethyl-3-chloro-4-methylbenzoylformate ≥97%
Ethyl-3-chloro-4-methylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₃
MW: 226.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801397
CAS Number: 951888-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR