Ethyl-2-methoxy-5-methylbenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-methoxy-5-methylphenyl)-2-oxoacetate

82463-500MG 82463-1G 82463-5G 82463-25G
77334-682EA 125.66 USD
77334-682 77344-080 77370-706 77382-812
Ethyl-2-methoxy-5-methylbenzoylformate ≥97%
Ethyl-2-methoxy-5-methylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801429
CAS Number: 859775-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR