Ethyl 2-oxo-2-(4-(trifluoromethoxy)phenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

82462-1G 82462-5G 82462-25G
77342-046EA 111.24 USD
77342-046 77370-704 77382-810
Ethyl 2-oxo-2-(4-(trifluoromethoxy)phenyl)acetate ≥95%
Ethyl 2-oxo-2-(4-(trifluoromethoxy)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₄
MW: 262.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16284944
CAS Number: 642459-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR