Ethyl-2,5-dichlorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2,5-dichlorophenyl)-2-oxoacetate

82459-500MG 82459-1G 82459-5G 82459-25G
77334-680EA 199.82 USD
77334-680 77358-874 77370-702 77391-346
Ethyl-2,5-dichlorobenzoylformate ≥95%
Ethyl-2,5-dichlorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801403
CAS Number: 951888-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR