Ethyl-3-chloro-5-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-chloro-5-fluorophenyl)-2-oxoacetate

82455-500MG 82455-1G 82455-5G 82455-25G
77334-678EA 151.41 USD
77334-678 77345-436 77370-700 77391-344
Ethyl-3-chloro-5-fluorobenzoylformate ≥95%
Ethyl 3-chloro-5-fluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₈ClFO₃
MW: 230.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201714
CAS Number: 845790-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR