Ethyl-4-chloro-3-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-chloro-3-methylphenyl)-2-oxoacetate

82454-500MG 82454-1G 82454-5G 82454-25G
77334-676EA 151.41 USD
77334-676 77345-434 77370-698 77391-342
Ethyl-4-chloro-3-methylbenzoylformate ≥95%
Ethyl 4-chloro-3-methylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₃
MW: 226.656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201715
CAS Number: 250642-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR