Ethyl-4-bromo-2-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-bromo-2-methylphenyl)-2-oxoacetate

82453-500MG 82453-1G 82453-5G 82453-25G
77334-674EA 130.81 USD
77334-674 77344-358 77370-696 77391-340
Ethyl-4-bromo-2-methylbenzoylformate ≥95%
Ethyl-4-bromo-2-methylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₃
MW: 271.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801390
CAS Number: 383363-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR