Ethyl-2-fluoro-4-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-fluoro-4-methylphenyl)-2-oxoacetate

82449-500MG 82449-1G 82449-5G 82449-25G
77334-672EA 211.15 USD
77334-672 77358-872 77370-694 77391-338
Ethyl-2-fluoro-4-methylbenzoylformate ≥95%
Ethyl-2-fluoro-4-methylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD09801423
CAS Number: 873547-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR