Ethyl 2-(5-fluoro-2-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82446-500MG 82446-1G 82446-5G 82446-25G
77334-668EA 125.66 USD
77334-668 77344-078 77370-690 77382-808
Ethyl 2-(5-fluoro-2-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(5-fluoro-2-methylphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MW: 210.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319624
CAS Number: 732251-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR