Ethyl 2-(3-bromo-2-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82443-500MG 82443-1G 82443-5G 82443-25G
77334-666EA 85.49 USD
77334-666 77342-258 77370-688 77382-806
Ethyl 2-(3-bromo-2-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-2-fluorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD23699501
CAS Number: 1426958-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR