Ethyl-3,5-dimethoxybenzoylformate ≥97%

Supplier: AOBChem USA

Warning

82442-500MG 82442-1G 82442-5G 82442-25G
77334-664EA 151.41 USD
77334-664 77345-406 77370-686 77391-334
Ethyl-3,5-dimethoxybenzoylformate ≥97%
Ethyl-3,5-dimethoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD09801406
CAS Number: 330551-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR