Ethyl-3-cyanobenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-cyanophenyl)-2-oxoacetate

82423-500MG 82423-1G 82423-5G 82423-25G
77334-660EA 217.33 USD
77334-660 77358-868 77370-682 77391-330
Ethyl-3-cyanobenzoylformate ≥97%
Ethyl-3-cyanobenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319613
CAS Number: 732249-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR