Ethyl-3-fluoro-4-methoxybenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-fluoro-4-methoxyphenyl)-2-oxoacetate

82422-500MG 82422-1G 82422-5G 82422-25G
77334-658EA 94.76 USD
77334-658 77342-532 77370-680 77382-804
Ethyl-3-fluoro-4-methoxybenzoylformate ≥95%
Ethyl 3-fluoro-4-methoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
MW: 226.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201699
CAS Number: 345-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR