Ethyl-2-naphthoyl formate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl (naphth-2-yl)(oxo)acetate, Ethyl 2-(naphthalen-2-yl)-2-oxoacetate

82421-500MG 82421-1G 82421-5G 82421-25G
77334-656EA 161.71 USD
77334-656 77346-020 77370-678 77391-328
Ethyl-2-naphthoyl formate ≥97%
Ethyl-2-naphthoyl formate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01934866
CAS Number: 73790-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR