Ethyl-4-fluoro-1-naphthoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-fluoronaphthalen-1-yl)-2-oxoacetate

82420-500MG 82420-1G 82420-5G 82420-25G
77334-654EA 234.84 USD
77334-654 77358-866 77370-676 77391-326
Ethyl-4-fluoro-1-naphthoylformate ≥97%
Ethyl-4-fluoro-1-naphthoylformate

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD09801427
CAS Number: 409081-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR