Ethyl -3,4-ethylenedioxybenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2-oxoacetate

82415-500MG 82415-1G 82415-5G 82415-25G
77334-650EA 134.93 USD
77334-650 77344-504 77370-672 77391-322
Ethyl -3,4-ethylenedioxybenzoylformate ≥95%
Ethyl -3,4-ethylenedioxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₅
MW: 236.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201701
CAS Number: 131030-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR