Ethyl-2-methoxybenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-methoxyphenyl)-2-oxoacetate

82413-500MG 82413-1G 82413-5G 82413-25G
77334-648EA 138.02 USD
77334-648 77344-656 77370-670 77391-320
Ethyl-2-methoxybenzoylformate ≥97%
Ethyl-2-methoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00858994
CAS Number: 66644-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR