Ethyl-3,5-dimethylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3,5-dimethylphenyl)-2-oxoacetate

82412-500MG 82412-1G 82412-5G 82412-25G
77334-646EA 86.52 USD
77334-646 77342-274 77370-668 77382-802
Ethyl-3,5-dimethylbenzoylformate ≥95%
Ethyl 3,5-dimethylbenzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₅
MW: 206.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311166
CAS Number: 100117-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR