Ethyl 2-oxo-2-(4-(trifluoromethyl)phenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

82399-1G 82399-5G 82399-25G
77340-920EA 62.83 USD
77340-920 77370-662 77382-798
Ethyl 2-oxo-2-(4-(trifluoromethyl)phenyl)acetate ≥95%
Ethyl 2-oxo-2-(4-(trifluoromethyl)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₃
MW: 246.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01934867
CAS Number: 73790-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR