1,2,3-Trichloro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

82370-500MG 82370-1G 82370-5G 82370-25G
77334-642EA 156.56 USD
77334-642 77345-632 77370-654 77391-316
1,2,3-Trichloro-5-iodobenzene ≥97%
1,2,3-Trichloro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₃I
Storage Temperature: Ambient
MDL Number: MFCD18400144
CAS Number: 64634-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR