Methyl 3-bromo-2-ethoxy-5-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

82338-500MG 82338-1G 82338-5G
77337-500EA 415.09 USD
77337-500 77358-864 77382-790
Methyl 3-bromo-2-ethoxy-5-isopropylbenzoate ≥97%
Methyl 3-bromo-2-ethoxy-5-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrO₃
MW: 301.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769275
CAS Number: 2901105-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR