2-Bromo-4-chloro-1-methyl-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

82337-500MG 82337-1G 82337-5G 82337-25G
77334-640EA 207.03 USD
77334-640 77358-862 77370-652 77391-314
2-Bromo-4-chloro-1-methyl-3-(methylsulfonyl)benzene ≥97%
2-Bromo-4-chloro-1-methyl-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO₂S
MW: 283.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769274
CAS Number: 2901065-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR