2-Bromo-4-chloro-3-(methoxymethyl)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

82334-500MG 82334-1G 82334-5G 82334-25G
77334-638EA 157.59 USD
77334-638 77345-684 77370-650 77391-312
2-Bromo-4-chloro-3-(methoxymethyl)-1-methylbenzene ≥97%
2-Bromo-4-chloro-3-(methoxymethyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769272
CAS Number: 2901098-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR