2-(Benzyloxy)-1-bromo-3-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

82319-500MG 82319-1G 82319-5G 82319-25G
77334-636EA 214.24 USD
77334-636 77358-860 77370-648 77391-310
2-(Benzyloxy)-1-bromo-3-methoxy-5-methylbenzene ≥97%
2-(Benzyloxy)-1-bromo-3-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
MW: 307.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769267
CAS Number: 2918829-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR