1-Bromo-2-isopropoxy-3-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

82318-500MG 82318-1G 82318-25G 82318-5G
77334-634EA 289.43 USD
77334-634 77358-858 77391-308 77382-788
1-Bromo-2-isopropoxy-3-methoxy-5-methylbenzene ≥97%
1-Bromo-2-isopropoxy-3-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769266
CAS Number: 2901106-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR