1-Bromo-2-ethoxy-3-methoxy-5-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
82317-500MG
82317-1G
82317-5G
77334-632EA
311.06
USD
77334-632
77358-856
77382-786
1-Bromo-2-ethoxy-3-methoxy-5-methylbenzene ≥97%
1-Bromo-2-ethoxy-3-methoxy-5-methylbenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrO₂ MW: 245.113 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34769265 CAS Number: 2385763-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...