3,4,5-Trichlorophenol ≥95%

Supplier: AOBChem USA

Warning

82314-1G 82314-5G 82314-25G
77340-918EA 62.83 USD
77340-918 77370-646 77382-784
3,4,5-Trichlorophenol ≥95%
3,4,5-Trichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130143
CAS Number: 609-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR