2-Bromo-1,3,4-trimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

82279-500MG 82279-1G 82279-5G 82279-25G
77334-628EA 187.46 USD
77334-628 77358-852 77370-642 77391-304
2-Bromo-1,3,4-trimethoxybenzene ≥97%
2-Bromo-1,3,4-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MW: 247.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043842
CAS Number: 25245-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR