1,4-Dichloro-2-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

82264-500MG 82264-1G 82264-5G
77337-020EA 404.79 USD
77337-020 77358-850 77382-774
1,4-Dichloro-2-iodo-5-methylbenzene ≥97%
1,4-Dichloro-2-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786703
CAS Number: 121020-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR