2-Bromo-6-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

82236-500MG 82236-1G 82236-25G 82236-5G
77334-618EA 284.28 USD
77334-618 77358-842 77391-294 77382-768
2-Bromo-6-fluoro-3-formylbenzoic acid ≥97%
2-Bromo-6-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrFO₃
MW: 247.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769235
CAS Number: 2383602-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR