5-Bromo-2,4-dichloroaniline ≥95%

Supplier: AOBChem USA

Warning

82111-1G 82111-5G 82111-25G
77340-832EA 60.77 USD
77340-832 77370-626 77382-764
5-Bromo-2,4-dichloroaniline ≥95%
5-Bromo-2,4-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrCl₂N
MW: 240.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226285
CAS Number: 258344-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR