Methyl 2-methoxy-5-(methoxymethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

82105-500MG 82105-1G 82105-5G
77335-736EA 363.59 USD
77335-736 77358-838 77382-762
Methyl 2-methoxy-5-(methoxymethyl)benzoate ≥97%
Methyl 2-methoxy-5-(methoxymethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769126
CAS Number: 91971-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR