3-Isopropoxy-2-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

82020-500MG 82020-1G 82020-5G
77335-980EA 371.83 USD
77335-980 77358-836 77382-760
3-Isopropoxy-2-methoxy-5-methylbenzaldehyde ≥97%
3-Isopropoxy-2-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34769065
CAS Number: 2901102-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR