1-Bromo-5-chloro-4-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

81831-500MG 81831-1G 81831-5G 81831-25G
77334-616EA 176.13 USD
77334-616 77346-566 77370-620 77391-292
1-Bromo-5-chloro-4-fluoro-2-methylbenzene ≥97%
1-Bromo-5-chloro-4-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817676
CAS Number: 1242339-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR