2,2,2-Trifluoro-1-(4-pentylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

81820-500MG 81820-1G 81820-5G 81820-25G
77334-614EA 89.61 USD
77334-614 77342-372 77370-618 77382-754
2,2,2-Trifluoro-1-(4-pentylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-pentylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₃O
MW: 244.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319982
CAS Number: 886369-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR