(2,4-Difluorophenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

81812-500MG 81812-1G 81812-5G 81812-25G
77334-612EA 161.71 USD
77334-612 77346-016 77370-614 77391-290
(2,4-Difluorophenyl)(ethyl)sulfane ≥97%
(2,4-Difluorophenyl)(ethyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂S
Storage Temperature: Ambient
MDL Number: MFCD30341004
CAS Number: 1084334-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR