2-Chloro-6-(cyclopropylmethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

81777-500MG 81777-1G 81777-5G 81777-25G
77334-608EA 182.31 USD
77334-608 77347-048 77370-608 77391-286
2-Chloro-6-(cyclopropylmethoxy)pyridine ≥97%
2-Chloro-6-(cyclopropylmethoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18367864
CAS Number: 108122-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR