4-Bromo-3,6-dichloro-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

81664-500MG 81664-1G 81664-5G 81664-25G
77334-606EA 231.75 USD
77334-606 77358-832 77370-606 77391-284
4-Bromo-3,6-dichloro-2-methylphenol ≥97%
4-Bromo-3,6-dichloro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34768911
CAS Number: 2378019-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR