3-Bromo-2,5-dichloro-6-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

81657-500MG 81657-1G 81657-5G 81657-25G
77334-604EA 161.71 USD
77334-604 77346-014 77370-604 77391-282
3-Bromo-2,5-dichloro-6-hydroxybenzoic acid ≥97%
3-Bromo-2,5-dichloro-6-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30297513
CAS Number: 1700077-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR