3-Bromo-2,5-dichloro-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81637-1G 81637-5G 81637-25G
77341-444EA 85.49 USD
77341-444 77370-602 77382-746
3-Bromo-2,5-dichloro-6-fluorobenzaldehyde ≥97%
3-Bromo-2,5-dichloro-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₂BrCl₂FO
MW: 271.899 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768902
CAS Number: 2901085-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR