(5-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

81443-500MG 81443-1G 81443-5G
77335-734EA 363.59 USD
77335-734 77358-822 77382-740
(5-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulfane ≥97%
(5-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34768804
CAS Number: 2901098-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR