1-Fluoro-2-isopropoxy-4-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

81403-500MG 81403-1G 81403-5G
77339-702EA 614.91 USD
77339-702 77358-816 77382-732
1-Fluoro-2-isopropoxy-4-(methoxymethyl)benzene ≥97%
1-Fluoro-2-isopropoxy-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂
MW: 198.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768789
CAS Number: 2901083-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR