2,5-Dimethyl-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81346-500MG 81346-1G 81346-25G 81346-5G
77334-596EA 300.76 USD
77334-596 77358-814 77391-276 77382-730
2,5-Dimethyl-4-propoxybenzaldehyde ≥97%
2,5-Dimethyl-4-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247273
CAS Number: 883532-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR