2-(Benzyloxy)-5-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81315-500MG 81315-1G 81315-5G
77339-208EA 566.5 USD
77339-208 77358-810 77382-726
2-(Benzyloxy)-5-fluoro-4-methylbenzaldehyde ≥97%
2-(Benzyloxy)-5-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34768746
CAS Number: 2901102-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR