5-Fluoro-2-isopropoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81307-500MG 81307-1G 81307-5G
77340-174EA 813.7 USD
77340-174 77358-806 77382-722
5-Fluoro-2-isopropoxy-4-methylbenzaldehyde ≥97%
5-Fluoro-2-isopropoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34768738
CAS Number: 2901095-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR