Ethyl 2-hydroxy-2-(4-methoxyphenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

81292-500MG 81292-1G 81292-5G 81292-25G
77334-588EA 242.05 USD
77334-588 77358-802 77370-586 77391-272
Ethyl 2-hydroxy-2-(4-methoxyphenyl)acetate ≥95%
Ethyl 2-hydroxy-2-(4-methoxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00972922
CAS Number: 36141-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR